missing translation for 'onlineSavingsMsg'
Learn More

VeriSpec™ Chlorite - ClO2- Standard for Ion Chromatography, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical

Catalog No. RV010910250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010910250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010910250 Supplier Ricca Chemical Company Supplier No. RV010910250C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Formulated for high performance and reliable results

  • 1000ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Bottle, Amber Glass
CAS 7758-19-2, 7732-18-5
CAS Min % 97.8726%, 0.1274%
CAS Max % 100%, 0.1326%
Specific Gravity 1.01
Physical Form Liquid
Molecular Formula ClNaO2
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
SMILES [O-]Cl=O.[Na+]
IUPAC Name sodium;chlorite
Molecular Weight (g/mol) 90.438
Quantity 250 mL
PubChem CID 23668197
ChEBI CHEBI:78667
Grade High Purity
Show More Show Less