missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chlorite, Anhydrous, Flakes, 77.5-82.5%, Spectrum™ Chemical

Catalog No. 18606436
Change view
Click to view available options
Quantity:
100 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606436 45 kg Amber Glass Bottle
18606432 100 g Amber Glass Bottle
18606434 2.5 kg Amber Glass Bottle
18606435 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606436 Supplier Spectrum Chemical Mfg Cor Supplier No. S124345KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1243, 7758-19-2, NaClO2

Spectrum™ Chemical Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis.

(CAS No. 7758-19-2, Molecular Formula: NaClO2) Sodium Chlorite, Anhydrous, Flakes

Chemical Identifiers

CAS 7758-19-2
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.44
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
IUPAC Name sodium chlorite
SMILES [Na+].[O-][Cl]=O

Specifications

CAS Max % 1
Physical Form Flakes or Powder
Quantity 45 kg
Percent Purity 77.5 to 82.5%
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Chlorite, Anhydrous, Flakes