missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, 0.0250 N (N/40), Ricca Chemical

Catalog No. R62820004C
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R62820004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R62820004C Supplier Ricca Chemical Company Supplier No. R62820004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Amber Glass Bottle
CAS 7758-05-6, 7681-11-0, 1310-73-2, 7732-18-5
Odor Odorless
Assay Percent Range 0.02500 ± 0.00001 N at 20°C
Physical Form Liquid
Molecular Formula IKO3
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Standard Solution
Type Analytical Volumetric Solution
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
Solvent or Matrix Water
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
Quantity 4 L
PubChem CID 23665710
Grade Laboratory
Chemical Name or Material Potassium Iodate-Iodide, 0.0250N (N/40)
Show More Show Less