missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™

Catalog No. LC196304
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC196304 4 L
LC196301 500 mL
LC196302 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC196304 Supplier LabChem Supplier No. LC196304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-05-6 , 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Color Colorless
CAS Max % 99.64, 0.36
Physical Form Liquid
Quantity 4 L
Linear Formula KIO3
Solubility Information Soluble in water
Formula Weight 214
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1000N ±0.0005N (0.0167M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Iodine vapor
Density 1g/mL
Chemical Name or Material Potassium Iodate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature