missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, 0.0500 N (N/20), Ricca Chemical

Catalog No. 627432
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
627432 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 627432 Supplier Ricca Chemical Company Supplier No. 627432

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7758-05-6 , 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0500N (N/20)
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Potassium Iodate