missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, 0.00564 N, 200 ppm as Cl2, Ricca Chemical

Catalog No. p-7239372
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
627016 500 mL Amber Glass Bottle
627032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 627016 Supplier Ricca Chemical Company Supplier No. 627016

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

APHA for Chlorine Testing

Chemical Identifiers

CAS 7758-05-6
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

CAS Min % 98.0, 0.02
CAS Max % 100.0, 0.02
Equivalence 1 mL = 0.2 mg Cl
Packaging Amber Glass Bottle
Quantity 500 mL
Concentration or Composition Notes 0.00564Normal
Chemical Name or Material Potassium Iodate