CAS RN 7664-41-7
Ammonia, ca. 7N solution in methanol
Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger | CAS: 7664-41-7 | H3N | 17.03 g/mol
Ammonium Hydroxide (26° Bé/NF/FCC), Fisher Chemical
CAS: 7664-41-7 Molecular Formula: H3N Molecular Weight (g/mol): 17.03 MDL Number: MFCD00011418 InChI Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N Synonym: ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol PubChem CID: 222 ChEBI: CHEBI:16134 IUPAC Name: ammonia SMILES: N
Ammonia, 0.5M solution in THF, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
Ammonia, ca. 7N solution in methanol, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
Ammonia, 2M in methanol
CAS: 7664-41-7 Molecular Formula: H3N Molecular Weight (g/mol): 17.03 MDL Number: MFCD00011418 InChI Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N Synonym: ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol PubChem CID: 222 ChEBI: CHEBI:16134 IUPAC Name: azane SMILES: N
Ammonia, 2M solution in methanol, AcroSeal™
CAS: 7664-41-7 | H3N | 17.03 g/mol
Ammonia, 7M in methanol
CAS: 7664-41-7 Molecular Formula: H3N Molecular Weight (g/mol): 17.03 MDL Number: MFCD00011418 InChI Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N Synonym: ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol PubChem CID: 222 ChEBI: CHEBI:16134 IUPAC Name: azane SMILES: N
Ammonia (ca. 4% in Methanol, ca. 2.0mol/L), TCI America™
CAS: 7664-41-7 Molecular Formula: H3N Molecular Weight (g/mol): 17.03 MDL Number: MFCD00011418 InChI Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N Synonym: ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol PubChem CID: 222 ChEBI: CHEBI:16134 IUPAC Name: ammonia SMILES: N
Ammonia (ca. 4% in Ethanol, ca. 2.0mol/L), TCI America™
CAS: 7664-41-7 Molecular Formula: H3N Molecular Weight (g/mol): 17.03 MDL Number: MFCD00011418 InChI Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N Synonym: ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol PubChem CID: 222 ChEBI: CHEBI:16134 IUPAC Name: ammonia SMILES: N
Ammonia (ca. 4% in Isopropyl Alcohol, ca. 2.0mol/L), TCI America™
CAS: 7664-41-7 Molecular Formula: H3N Molecular Weight (g/mol): 17.03 MDL Number: MFCD00011418 InChI Key: QGZKDVFQNNGYKY-UHFFFAOYSA-N Synonym: ammonia,ammonia gas,spirit of hartshorn,nitro-sil,ammoniakgas,ammonia anhydrous,anhydrous ammonia,ammonia, anhydrous,ammoniak,am-fol PubChem CID: 222 ChEBI: CHEBI:16134 IUPAC Name: ammonia SMILES: N
Ammonia-Ammonium Chloride Buffer TS, (U.S.P. Test Solution), Spectrum™ Chemical
A-266, 12125-02-9