missing translation for 'onlineSavingsMsg'
Learn More

Ammonia (ca. 4% in Isopropyl Alcohol, ca. 2.0mol/L), TCI America™

Catalog No. A2237500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2237500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2237500ML Supplier TCI America Supplier No. A2237500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name ammonia
SMILES N

Specifications

Color Colorless
UN Number 1219
Quantity 500 mL
Formula Weight 17.03
Physical Form Liquid
Chemical Name or Material Ammonia (ca. 4% in Isopropyl Alcohol, ca. 2.0mol/L)
EINECSNumber (1)-0391
RTECSNumber BO0875000
TSCA Yes
Recommended Storage Refrigerator