missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 7M in methanol

Catalog No. AAH30382AU
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH30382AU 1000 mL
AAH30382AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH30382AU Supplier Thermo Scientific Chemicals Supplier No. H30382AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7664-41-7 | H3N | 17.03 g/mol

Microwave-assisted synthesis of HX salts of primary amines from their corresponding halides and 7 M ammonia in methanol provides practical high yield access to even volatile primary amines for parallel synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name azane
SMILES N

Specifications

Linear Formula NH3
UN Number UN3286
Concentration 7M in methanol
Density 0.779
Merck Index 14,492
Solubility Information Fully misicible in water.
Formula Weight 17.03
Quantity 1000 mL
Flash Point 14°C (57°F)
Sensitivity Hygroscopic
Chemical Name or Material Ammonia
Show More Show Less
Hazard Category H225-H301+H311+H331-H314-H370
Hazard Statement GHS H Statement
H225-H311-H331-H370-H314
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 231-635-3
RTECSNumber BO0875000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only