accessibility menu, dialog, popup

UserName

BNOCH3HOOH

CAS RN 762262-09-9

IUPAC Name: (2-methoxypyridin-4-yl)boronic acid
Synonyms: 2-methoxypyridin-4-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: DHQMUJSACXTPEA-UHFFFAOYSA-N
SMILES: COC1=NC=CC(=C1)B(O)O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)