missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-4-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M27301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27301G Supplier TCI America Supplier No. M27301G

Specifications

Chemical Name or Material 2-Methoxypyridine-4-boronic Acid (contains varying amounts of Anhydride)
CAS 762262-09-9
Molecular Formula C6H8BNO3
Quantity 1 g
MDL Number MFCD07368877
Synonym 2-methoxypyridine-4-boronic acid, 2-methoxypyridin-4-yl boronic acid, 2-methoxypyridne-4-boronic acid, 2-methoxy-4-pyridineboronic acid, 2-methoxypyridin-4-yl-4-boronic acid, 2-methoxy-4-pyridyl boronic acid, boronic acid, 2-methoxy-4-pyridinyl, 2-methoxypyridne-4-boronicacid, pubchem17079, acmc-209p2b
InChI Key DHQMUJSACXTPEA-UHFFFAOYSA-N
SMILES COC1=NC=CC(=C1)B(O)O
IUPAC Name (2-methoxypyridin-4-yl)boronic acid
Molecular Weight (g/mol) 152.94
PubChem CID 23546919
Formula Weight 152.94
Physical Form Crystalline Powder
Show More Show Less