missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-4-boronic acid, 95%

Catalog No. AC454372500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454372500 250 mg
AC454370010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454372500 Supplier Thermo Scientific Chemicals Supplier No. 454372500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 762262-09-9 | C6H8BNO3 | 152.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 762262-09-9
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD07368877
InChI Key DHQMUJSACXTPEA-UHFFFAOYSA-N
Synonym 2-methoxypyridine-4-boronic acid, 2-methoxypyridin-4-yl boronic acid, 2-methoxypyridne-4-boronic acid, 2-methoxy-4-pyridineboronic acid, 2-methoxypyridin-4-yl-4-boronic acid, 2-methoxy-4-pyridyl boronic acid, boronic acid, 2-methoxy-4-pyridinyl, 2-methoxypyridne-4-boronicacid, pubchem17079, acmc-209p2b
PubChem CID 23546919
IUPAC Name (2-methoxypyridin-4-yl)boronic acid
SMILES COC1=NC=CC(=C1)B(O)O

Specifications

Melting Point 65.0°C to 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 152.95
Percent Purity 95%
Chemical Name or Material 2-Methoxypyridine-4-boronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only