accessibility menu, dialog, popup

UserName

CAS RN 762-21-0

CH3OOOOH3C

CAS RN 762-21-0

IUPAC Name: 1,4-diethyl but-2-ynedioate
Synonyms: 1,4-diethyl but-2-ynedioate
Molecular Weight (g/mol): 170.16
Molecular Formula: C8H10O4
InChi Key: STRNXFOUBFLVIN-UHFFFAOYSA-N
SMILES: CCOC(=O)C#CC(=O)OCC
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)