missing translation for 'onlineSavingsMsg'
Learn More

Diethyl acetylenedicarboxylate, 96%

Catalog No. AAA1171936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1171936 500 g
AAA1171914 25 g
AAA1171922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1171936 Supplier Thermo Scientific Chemicals Supplier No. A1171936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 762-21-0 | C8H10O4 | 170.164 g/mol

Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl acetylenedicarboxylate is used as a protein cross-linker. It is also used in the synthesis of 3,4,5-trisubstituted 2(5H)-furanone derivatives, highly functionalized thiazolidinone derivatives, novel cyclic peroxide glucosides and 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction.

Solubility
Soluble in ethanol, ethyl ether, CCl4, Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with strong acids and strong bases.

Chemical Identifiers

CAS 762-21-0
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00009186
InChI Key STRNXFOUBFLVIN-UHFFFAOYSA-N
Synonym diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate
PubChem CID 69803
IUPAC Name diethyl but-2-ynedioate
SMILES CCOC(=O)C#CC(=O)OCC

Specifications

Melting Point 1°C to 2°C
Density 1.067
Boiling Point 108°C to 110°C (11 mmHg)
Flash Point 94°C (201°F)
Refractive Index 1.443
Quantity 500 g
UN Number UN1760
Beilstein 743166
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,ethyl ether,CCl4,Insoluble in water.
Formula Weight 170.16
Percent Purity 96%
Chemical Name or Material Diethyl acetylenedicarboxylate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 212-095-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only