missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Acetylenedicarboxylate 96.0+%, TCI America™

Catalog No. A008925ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A008925ML 25 mL
A0089250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A008925ML Supplier TCI America Supplier No. A008925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 762-21-0
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00009186
InChI Key STRNXFOUBFLVIN-UHFFFAOYSA-N
Synonym diethyl acetylenedicarboxylate, 2-butynedioic acid, diethyl ester, diethyl 2-butynedioate, 1,4-diethyl but-2-ynedioate, acetylenedicarboxylic acid, diethyl ester, acetylenedicarboxylic acid diethyl ester, diethylacetylenedicarboxylate, bis-ethoxycarbonyl acetylene, diethyl acetylene dicarboxylate, di-ethyl acetylenedicarboxylate
PubChem CID 69803
IUPAC Name diethyl but-2-ynedioate
SMILES CCOC(=O)C#CC(=O)OCC

Specifications

Melting Point 1°C
Color Yellow
Boiling Point 115°C
Quantity 25 mL
UN Number 1760
Formula Weight 170.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Acetylenedicarboxylate