accessibility menu, dialog, popup

UserName

CH3CH3HOCH3CH3HO

CAS RN 76-09-5

IUPAC Name: 2,3-dimethylbutane-2,3-diol
Synonyms: pinacol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: IVDFJHOHABJVEH-UHFFFAOYSA-N
SMILES: CC(C)(O)C(C)(C)O
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)