missing translation for 'onlineSavingsMsg'
Learn More

Pinacol, 99%

Catalog No. AAB2500636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2500636 500 g
AAB2500614 25 g
AAB2500622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2500636 Supplier Thermo Scientific Chemicals Supplier No. B2500636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76-09-5 | C6H14O2 | 118.176 g/mol

Pinacol is used in the preparation of synthetic intermediates like pinacolborane, bis(pinacolato)diboron and pinacolchloroborane by reaction with boron trichloride. Further, it plays an important role as an intermediate for biologically active compounds like antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacol is used in the preparation of synthetic intermediates like pinacolborane, bis(pinacolato)diboron and pinacolchloroborane by reaction with boron trichloride. Further, it plays an important role as an intermediate for biologically active compounds like antiviral, antibacterial, antifungal and antituberculous.

Solubility
Soluble in hot water, alcohol, and diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases and strong acids.

Chemical Identifiers

CAS 76-09-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004462
InChI Key IVDFJHOHABJVEH-UHFFFAOYSA-N
Synonym pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane
PubChem CID 6425
IUPAC Name 2,3-dimethylbutane-2,3-diol
SMILES CC(C)(C(C)(C)O)O

Specifications

Melting Point 42°C to 45°C
Boiling Point 171°C to 172°C
Flash Point 77°C (170°F)
Quantity 500 g
Beilstein 1340501
Sensitivity Hygroscopic
Merck Index 14,7439
Solubility Information Soluble in hot water,alcohol,and diethyl ether.
Formula Weight 118.18
Percent Purity 99%
Chemical Name or Material Pinacol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 200-933-5
RTECSNumber EK1720000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only