missing translation for 'onlineSavingsMsg'
Learn More

Buffer, Reference Standard, pH 11.00 ± 0.01 at 25°C, Ricca Chemical

Catalog No. 16101
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Clear PET-G Bottle
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
16101 4 L Natural Poly Bottle
R161016 500 mL Clear PET-G Bottle
161032 1 L Natural Poly Bottle
16105 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16101 Supplier Ricca Chemical Company Supplier No. RI16101
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Buffer, Reference Standard
Name Note pH 11.00± 0.01 at 25°C
Melting Point 0°C
CAS 7558-79-4, 1310-73-2, 7732-18-5
Boiling Point 100°C
CAS Min % 97.0, 1.02, 0.11
Color Colorless
CAS Max % 100.0, 1.06, 0.11
Concentration 1X
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 11
Physical Form Liquid
Quantity 4 L
Solution Type pH Buffer
Packaging Natural Poly Bottle
Sterility Non-sterile
Solubility Information Miscible
Grade Reference Standard
Show More Show Less