missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 7, Phosphate buffer

Catalog No. AC258595000
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC258595000 500 mL
AC258591000 100 mL
AC258590010 1 L
AC258590025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC258595000 Supplier Thermo Scientific Chemicals Supplier No. 258595000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7558-79-4 | H2KO4P

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer

Specifications

Chemical Name or Material Buffer solution pH 7
CAS 7558-79-4, 7732-18-5
Density 1.0030g/mL
Quantity 500 mL
Molecular Formula H2KO4P
MDL Number MFCD03097944
Packaging Plastic bottle
Specific Gravity 1.003
Merck Index 14, 7659
Vapor Pressure 14mmHg at 20°C
Solubility Information Solubility in water: soluble.
Formula Weight 136.09
Show More Show Less
What is the concentration of the Buffer solution pH 7, Phosphate buffer?

The Buffer solution pH 7, Phosphate buffer has a concentration of 0.1 M.

What is the composition of the Buffer solution pH 7, Phosphate buffer?

The composition of the Buffer solution pH 7, Phosphate buffer is 3.54 g potassium dihydrogen phosphate and 14.7 g di-sodium hydrogen phosphate per liter.

RUO – Research Use Only