CAS RN 7440-41-7
Beryllium foil, 0.05mm (0.002in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium lump, irregular, 3-38mm (0.1-1.5in), 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.25mm (0.01in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.1mm (0.004in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium powder, -325 mesh, 99+% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.1mm (0.004in) thick, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium wire, 0.125mm (0.005 in.) dia., annealed, 99.7% (metals basis), Thermo Scientific™
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.5mm (0.02in) thick, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium AA Standard, 1000 ppm Be in 3% HNO3, Ricca Chemical
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium ICP-MS Standard, 1000 ppm Be in 3% HNO3, Ricca Chemical
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium Organometallic Standards, SPEX CertiPrep™
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Analytes C Interference Check Standard For ICP/MS, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Beryllium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
- 1
- 2