missing translation for 'onlineSavingsMsg'
Learn More

Beryllium AA Standard, 1000 ppm Be in 3% HNO3, Ricca Chemical

Catalog No. ABE1KN500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ABE1KN500 500 mL Natural Poly Bottle
ABE1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE1KN500 Supplier Ricca Chemical Company Supplier No. ABE1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-41-7 , 7697-37-2 , 7440-41-7
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1,000ppm Be
CAS Min % 88.0, 9.91, 0.09
CAS Max % 92.0, 10.32, 0.09
Equivalence 1 mL = 1 mg Be
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Quantity 500 mL
Chemical Name or Material Beryllium AA Standard
Show More Show Less