missing translation for 'onlineSavingsMsg'
Learn More

Beryllium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™

Catalog No. CLBE21BY
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLBE21BY 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLBE21BY Supplier SPEX CertiPrep Group LLC Supplier No. CLBE21BY

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-41-7 , 7697-37-2 , 7732-18-5
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]

Specifications

CAS Max % 0.0, 2.0, 100.0
Analyte or Component Names Beryllium Standard
Physical Form Fluid
Element Beryllium Standard
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Beryllium Standard for ICP/MS - Claritas PPT Grade, ICP/MS
Recommended Storage Ambient