CAS RN 7440-38-2
Arsenic lump, 99.999% (metals basis)
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic powder, -20 mesh, 99% (metals basis)
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic, plasma standard solution, Specpure™ As 1000μg/mL
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
CAS: 7440-38-2 | As | 74.92 g/mol
Arsenic ICP Standard, 1000 ppm As in 3% HCl, Ricca Chemical
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic ICP Standard, 10,000 ppm As in 5% HNO3, Ricca Chemical
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic ICP Standard, 1000 ppm As in 3% HNO3, Ricca Chemical
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic ICP-MS Standard, 1000 ppm As in 3% HNO3, Ricca Chemical
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Standard, 1000μg/g in Base Oil 75, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Arsenic Standard for ICP/MS, 2% HNO3, SPEX CertiPrep™
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
- 1
- 2