missing translation for 'onlineSavingsMsg'
Learn More

Arsenic ICP Standard, 1000 ppm As in 3% HNO3, Ricca Chemical

Catalog No. PAS1KN500
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PAS1KN500 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PAS1KN500 Supplier Ricca Chemical Company Supplier No. PAS1KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 7440-38-2, 7697-37-2, 7440-38-2, 7732-18-5
CAS Min % 90.0, 7.47, 0.09
Concentration or Composition Notes 1,000ppm As
CAS Max % 94.0, 7.78, 0.09
Equivalence 1 mL = 1 mg As
Molecular Formula As
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
SMILES [As]
IUPAC Name arsenic
Molecular Weight (g/mol) 74.92
PubChem CID 5359596
ChEBI CHEBI:27563
Chemical Name or Material Arsenic ICP Standard
Show More Show Less