missing translation for 'onlineSavingsMsg'
Learn More

Arsenic lump, 99.999% (metals basis)

Catalog No. AA0059022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0059022 100 g
AA0059014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0059022 Supplier Thermo Scientific Chemicals Supplier No. 00059022

missing translation for 'validMainframeMsg'

CAS: 7440-38-2 | As | 74.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-38-2
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
Synonym arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Chemical Name or Material Arsenic lump
Hazard Category H301+H331-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 213-148-6
RTECSNumber CG0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only