missing translation for 'onlineSavingsMsg'
Learn More

Arsenic ICP Standard, 10,000 ppm As in 5% HNO3, Ricca Chemical

Catalog No. p-3251322
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PAS10KN50 Natural Poly Bottle
PAS10KN100 Natural LDPE Bottle
PAS10KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PAS10KN50 Supplier Ricca Chemical Company Supplier No. PAS10KN50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-38-2 , 7697-37-2 , 7440-38-2 , 7732-18-5
Molecular Formula As
Molecular Weight (g/mol) 74.92
MDL Number MFCD00085309
InChI Key RQNWIZPPADIBDY-UHFFFAOYSA-N
PubChem CID 5359596
ChEBI CHEBI:27563
IUPAC Name arsenic
SMILES [As]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm As
CAS Min % 67.0, 30.24, 0.67
CAS Max % 70.0, 31.48, 0.7
Equivalence 1 mL = 10 mg As
Chemical Name or Material Arsenic ICP Standard