accessibility menu, dialog, popup

UserName

H3C(S)OHONH2CH3

CAS RN 72-18-4

IUPAC Name: (2S)-2-amino-3-methylbutanoic acid
Synonyms: L-valine|Val|L-valin|2-amino-3-methylbutyric acid|L-(+)-a-aminoisovaleric acid
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: KZSNJWFQEVHDMF-BYPYZUCNSA-N
SMILES: CC(C)[C@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (19)
Quantity 
  • (3)
  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
Grade 
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (7)
  • (2)
  • (2)
  • (5)
  • (3)
  • (11)
  • (2)
  • (19)
  • (3)
  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)