missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 117.15 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Valine |
Safety and Handling
| EINECSNumber | (9)-1604 |
| RTECSNumber | YV9361000 |
| TSCA | Yes |