missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015350
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015350 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015350 Supplier Merck Emd Millipore Supplier No. PHR11721G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Valine is an essential amino acid, involved in the biosynthesis of glutamine and alanine. Valine being a branched-chain amino acid (BCAA), maintains a balance among the BCAAs. L-Valine serves as an energy fuel.

Specifications

CAS 72-18-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (CH3)2 CHCH(NH2)CO2 H
MDL Number MFCD00064220
Melting Point 295°C to 300°C (sublimation)
Synonym L-Valine; (S)-alpha-Aminoisovaleric acid; L-2-Amino-3-methylbutanoic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 117.15
Grade Certified Reference Material
Show More Show Less