Learn More
Thermo Scientific Chemicals L-Valine, 99%
Description
L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.
Solubility
Soluble in water. Slightly soluble in methanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Spécifications
Spécifications
| Melting Point | ∼315°C |
| Assay Percent Range | 99% |
| Beilstein | 1721136 |
| Merck Index | 14,9909 |
| Solubility Information | Soluble in water. Slightly soluble in methanol and acetone. |
| Density | 1.23 |
| Quantity | 500 g |
| Odor | Odorless |
| Formula Weight | 117.15 |
| Optical Rotation | +27° (c=5 in 5N HCl) |
| Afficher plus de résultats |
Sécurité et manipulation
| EINECSNumber | 200-773-6 |
| RTECSNumber | YV9361000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only