missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ L-Valine, MilliporeSigma™ Supelco™

Catalog No. 111015349
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015349 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015349 Supplier Merck Emd Millipore Supplier No. 50848100MG
Only null left
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

CAS 72-18-4
Product Line TraceCERT
Physical Form Neat
Linear Formula (CH3)2 CHCH(NH2)CO2 H
MDL Number MFCD00064220
Melting Point 295°C to 300°C (sublimation)
Synonym (S)-alpha-Aminoisovaleric acid; L-2-Amino-3-methylbutanoic acid
Type Certified Reference Material
Quantity 100 mg
Formula Weight 117.15
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC