accessibility menu, dialog, popup

UserName

H2OH2OH2OCH3CH3S(R)(R)NHO(R)NH2ON(S)HOO

CAS RN 7177-48-2

IUPAC Name: (2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate
Synonyms: principen campicillin aminobenzylpenicillin trihydrate ampicillin trihydrate penbritin
Molecular Weight (g/mol): 403.45
Molecular Formula: C16H25N3O7S
InChi Key: RXDALBZNGVATNY-CWLIKTDRSA-N
SMILES: O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)