missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ampicillin trihydrate

Catalog No. AAJ6651414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6651414 25 g
AAJ6651436 500 g
AAJ6651422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6651414 Supplier Thermo Scientific Chemicals Supplier No. J6651414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commonly used to select for ampicillin resistance in mutated and transformed cells

Commonly used to select for ampicillin resistance in mutated and transformed cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 7177-48-2
Chemical Name or Material Ampicillin trihydrate
Color White
Content And Storage Keep cold
Form Powder
Product Type Ampicillin Trihydrate
Quantity 25 g
Molecular Weight (g/mol) 403.45
Solubility Soluble in water. Practically insoluble in ethanol

RUO – Research Use Only