missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Trihydrate 98.0+%, TCI America™

Catalog No. A20925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20925G 5 g
A209225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20925G Supplier TCI America Supplier No. A20925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7177-48-2
Molecular Formula C16H25N3O7S
Molecular Weight (g/mol) 403.45
MDL Number MFCD00072036
InChI Key RXDALBZNGVATNY-CWLIKTDRSA-N
Synonym ampicillin trihydrate, aminobenzylpenicillin trihydrate, amperil, ampinova, trafarbiot, amcap, cymbi, ampicillin 3-wasser, unii-hxq6a1n7r6, alpha-aminobenzylpenicillin trihydrate
PubChem CID 23565
ChEBI CHEBI:31209
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;trihydrate
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.O.O.O

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 5 g
Formula Weight 349.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ampicillin Trihydrate
RTECSNumber XH8425000
TSCA No
Recommended Storage Refrigerator