CAS RN 70955-01-0
Molecular sieves, 4A, 1-2mm (0.04-0.08in) dia. pellets
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves 4A, 8 to 12 mesh
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves 4A, Cylinders, 3.2mm diameter
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves, 4A, 3-4mm (0.12-0.16in) dia. pellets
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves 4A, 10 to 18 mesh
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: Molecular sieves SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves, 4A, -8+12 (ca 2mm) beads
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves, 4A, powder
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves 4A, 4 to 8 mesh
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves, 4A, 0.4-0.8mm (0.02-0.03in) beads
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular sieves 4A, powder <50 micron
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular Sieves, 4A, Pellets, Dia.: 3.2mm, Solstice
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular Sieves, 4 Å, 8 to 12mesh, Solstice
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC
Molecular Sieves, 4 Å, 4 to 8mesh, Solstice
CAS: 70955-01-0 Molecular Formula: C20H25FN2O8 Molecular Weight (g/mol): 440.42 MDL Number: MFCD00131613 InChI Key: FJUZEZRZKVMAMD-UHFFFAOYNA-N PubChem CID: 73906282 IUPAC Name: (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid SMILES: COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC