missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, 10 to 18 mesh

Catalog No. AC264755000
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264755000 500 g Glass bottle
AC264750050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC264755000 Supplier Thermo Scientific Chemicals Supplier No. 264755000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name Molecular sieves
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Beige
Quantity 500 g
Bulk Density 680 to 760g/L
Water Adsorption > 20% (50% rH, 20°C, 24 h)
Residual Water < 1.5% (550°C, 2 hrs.)
Packaging Glass bottle
Physical Form Granules
Chemical Name or Material Molecular sieves 4A
Hazard Category HYGROSCOPIC

RUO – Research Use Only