accessibility menu, dialog, popup

UserName

CAS RN 6971-51-3

IUPAC Name: (3-methoxyphenyl)methanol
Synonyms: 3-methoxybenzyl alcohol 3-anise alcohol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: IIGNZLVHOZEOPV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CO)=C1
Boiling Point 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)