missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl Alcohol 97.0+%, TCI America™

Catalog No. M110125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M110125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M110125G Supplier TCI America Supplier No. M110125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6971-51-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004637
InChI Key IIGNZLVHOZEOPV-UHFFFAOYSA-N
Synonym 3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol
PubChem CID 81437
ChEBI CHEBI:1580
IUPAC Name (3-methoxyphenyl)methanol
SMILES COC1=CC=CC(=C1)CO

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzyl Alcohol