missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl alcohol, 98%

Catalog No. AC125770250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125770250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC125770250 Supplier Thermo Scientific Chemicals Supplier No. 125770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6971-51-3 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6971-51-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004637
InChI Key IIGNZLVHOZEOPV-UHFFFAOYSA-N
Synonym 3-methoxybenzyl alcohol, 3-methoxyphenyl methanol, m-methoxybenzyl alcohol, 3-methoxybenzylalcohol, m-anisyl alcohol, 3-anise alcohol, 3-anisyl alcohol, benzenemethanol, 3-methoxy, unii-i1xc3698n5, 3-methoxyphenyl methan-1-ol
PubChem CID 81437
ChEBI CHEBI:1580
IUPAC Name (3-methoxyphenyl)methanol
SMILES COC1=CC=CC(=C1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown to Yellow
Density 1.1100g/mL
Boiling Point 250.0°C (723.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5425 to 1.5445
Linear Formula CH3OC6H4CH2OH
Quantity 25 mL
Beilstein 06, 896
Specific Gravity 1.11
Solubility Information Solubility in water: immiscible
Formula Weight 138.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzyl alcohol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 230-200-5

RUO – Research Use Only