accessibility menu, dialog, popup

UserName

CH3OOHOCH3

CAS RN 6880-04-2

IUPAC Name: 4-methoxy-3-methylbenzoic acid
Synonyms: 4-methoxy-3-methylbenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DNMUMZLKDOZMEY-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results