missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. M27285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27285G 5 g
M27281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27285G Supplier TCI America Supplier No. M27285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6880-04-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00270114
InChI Key DNMUMZLKDOZMEY-UHFFFAOYSA-N
Synonym 3-methyl-4-methoxybenzoic acid, 4-methoxy-3-methyl-benzoic acid, 4-methoxy-3-methylbenzoicacid, 3-methyl-p-anisic acid, 4-methoxy-m-toluic acid, benzoic acid, 4-methoxy-3-methyl, ksc495k5j, 4-me-thoxy-3-methylbenzoic acid, 4-methoxy-3-methyl benzoic acid
PubChem CID 2759583
IUPAC Name 4-methoxy-3-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)C(=O)O)OC

Specifications

Melting Point 197°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-methylbenzoic Acid