accessibility menu, dialog, popup

UserName

CAS RN 67881-98-5

H3CCH2OOOPOOONCH3CH3H3C

CAS RN 67881-98-5

IUPAC Name: trimethyl[2-({2-[(2-methylprop-2-enoyl)oxy]ethyl phosphonato}oxy)ethyl]azanium
Synonyms: polyphosphorylcholine glycol acrylate
Molecular Weight (g/mol): 295.27
Molecular Formula: C11H22NO6P
InChi Key: ZSZRUEAFVQITHH-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCOP([O-])(=O)OCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)