missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Methacryloyloxyethyl phosphorylcholine

Catalog No. 7301145G
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7301145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7301145G Supplier Sigma Aldrich Supplier No. 7301145G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methacryloyloxyethyl phosphorylcholine

Specifications

CAS 67881-98-5
Melting Point 143°C to 148°C
Molecular Formula C11H22NO6P
Linear Formula C11H22NO6P
MDL Number MFCD11112180
Quantity 5 g
Synonym MPC
Molecular Weight (g/mol) 295.27
Percent Purity 97%