missing translation for 'onlineSavingsMsg'
Learn More

2-(Methacryloyloxy)ethyl 2-(Trimethylammonio)ethyl Phosphate 96.0+%, TCI America™

Catalog No. M20055G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20055G 5 g
M20051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20055G Supplier TCI America Supplier No. M20055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67881-98-5
Molecular Formula C11H22NO6P
Molecular Weight (g/mol) 295.272
InChI Key ZSZRUEAFVQITHH-UHFFFAOYSA-N
Synonym Phosphoric Acid 2-(Methacryloyloxy)ethyl 2-(Trimethylammonio)ethyl Ester, 2-Methacryloyloxyethyl Phosphorylcholine
PubChem CID 128934
IUPAC Name 2-(2-methylprop-2-enoyloxy)ethyl 2-(trimethylazaniumyl)ethyl phosphate
SMILES CC(=C)C(=O)OCCOP(=O)([O-])OCC[N+](C)(C)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 295.27
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Methacryloyloxy)ethyl 2-(Trimethylammonio)ethyl Phosphate
EINECSNumber (2)-3871
TSCA No
Recommended Storage Freezer