accessibility menu, dialog, popup

UserName

CAS RN 6728-31-0

CH3O(E)

CAS RN 6728-31-0

IUPAC Name: (4E)-hept-4-enal
Synonyms: cis-4-heptenal
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: VVGOCOMZRGWHPI-ONEGZZNKSA-N
SMILES: CC\C=C\CCC=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)