missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Heptenal, 96%

Catalog No. AC298220100
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC298220100 10 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC298220100 Supplier Thermo Scientific Chemicals Supplier No. 298220100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6728-31-0 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6728-31-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00014682
InChI Key VVGOCOMZRGWHPI-ARJAWSKDSA-N
Synonym cis-4-heptenal, cis-4-hepten-1-al, z-hept-4-enal, z-4-heptenal, 4-heptenal, z, z-4-hepten-1-al, hept-cis-4-enal, unii-5bj99wwp64, 4-z-heptenal, hept-4 z-enal
PubChem CID 5362814
IUPAC Name (Z)-hept-4-enal
SMILES CCC=CCCC=O

Specifications

Color Colorless to Yellow
Density 0.8490g/mL
Boiling Point 46.0°C (13.0 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4320 to 1.4360
Linear Formula CH3CH2CH=CHCH2CH2CHO
Quantity 10 mL
Specific Gravity 0.849
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,propylene glycol
Formula Weight 112.17
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material cis-4-Heptenal, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 229-779-7

RUO – Research Use Only