CAS RN 670-54-2
Tetracyanoethylene, 98%
CAS: 670-54-2 Molecular Formula: C6N4 Molecular Weight (g/mol): 128.094 MDL Number: MFCD00001850 InChI Key: NLDYACGHTUPAQU-UHFFFAOYSA-N Synonym: tetracyanoethylene,tetracyanoethene,ethylenetetracarbonitrile,tcne,ethenetetracarbonitrile,ethene, tetracyano,1,1,2,2-ethenetetracarbonitrile,tetrakyanethylen,1,1,2,2-tetracyanoethene,1,1,2,2-tetracyanoethylene PubChem CID: 12635 IUPAC Name: ethene-1,1,2,2-tetracarbonitrile SMILES: C(#N)C(=C(C#N)C#N)C#N
Tetracyanoethylene, 98%, Thermo Scientific™
CAS: 670-54-2 Molecular Formula: C6N4 Molecular Weight (g/mol): 128.09 MDL Number: 00001850 InChI Key: NLDYACGHTUPAQU-UHFFFAOYSA-N Synonym: TCNE IUPAC Name: eth-1-ene-1,1,2,2-tetracarbonitrile SMILES: N#CC(C#N)=C(C#N)C#N
Tetracyanoethylene, 98%
CAS: 670-54-2 Molecular Formula: C6N4 Molecular Weight (g/mol): 128.09 InChI Key: NLDYACGHTUPAQU-UHFFFAOYSA-N Synonym: tetracyanoethylene,tetracyanoethene,ethylenetetracarbonitrile,tcne,ethenetetracarbonitrile,ethene, tetracyano,1,1,2,2-ethenetetracarbonitrile,tetrakyanethylen,1,1,2,2-tetracyanoethene,1,1,2,2-tetracyanoethylene PubChem CID: 12635 IUPAC Name: ethene-1,1,2,2-tetracarbonitrile SMILES: C(#N)C(=C(C#N)C#N)C#N
Tetracyanoethylene (purified by sublimation) 98.0+%, TCI America™
CAS: 670-54-2 Molecular Formula: C6N4 Molecular Weight (g/mol): 128.094 MDL Number: MFCD00001850 InChI Key: NLDYACGHTUPAQU-UHFFFAOYSA-N Synonym: tetracyanoethylene,tetracyanoethene,ethylenetetracarbonitrile,tcne,ethenetetracarbonitrile,ethene, tetracyano,1,1,2,2-ethenetetracarbonitrile,tetrakyanethylen,1,1,2,2-tetracyanoethene,1,1,2,2-tetracyanoethylene PubChem CID: 12635 IUPAC Name: ethene-1,1,2,2-tetracarbonitrile SMILES: C(#N)C(=C(C#N)C#N)C#N
Tetracyanoethylene 98.0+%, TCI America™
CAS: 670-54-2 Molecular Formula: C6N4 Molecular Weight (g/mol): 128.094 MDL Number: MFCD00001850 InChI Key: NLDYACGHTUPAQU-UHFFFAOYSA-N Synonym: tetracyanoethylene,tetracyanoethene,ethylenetetracarbonitrile,tcne,ethenetetracarbonitrile,ethene, tetracyano,1,1,2,2-ethenetetracarbonitrile,tetrakyanethylen,1,1,2,2-tetracyanoethene,1,1,2,2-tetracyanoethylene PubChem CID: 12635 IUPAC Name: ethene-1,1,2,2-tetracarbonitrile SMILES: C(#N)C(=C(C#N)C#N)C#N