missing translation for 'onlineSavingsMsg'
Learn More

Tetracyanoethylene, 98%, Thermo Scientific™

Catalog No. AC138050050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC138050050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC138050050 Supplier Thermo Scientific Chemicals Supplier No. 138050050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 670-54-2
Molecular Formula C6N4
Molecular Weight (g/mol) 128.09
MDL Number 00001850
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
Synonym TCNE
IUPAC Name eth-1-ene-1,1,2,2-tetracarbonitrile
SMILES N#CC(C#N)=C(C#N)C#N

Specifications

Melting Point 196°C to 201°C
Boiling Point 223°C (760 mmHg)
Infrared Spectrum Conforms to structure
Assay Percent Range ≥ 97.5% (Ex N)
Packaging Glass Bottle
Linear Formula (NC)2C=C(CN)2
Quantity 5 g
UN Number 3439
Sensitivity Moisture sensitive
Fieser 13-289
Merck Index 9340
Formula Weight 128.09
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Tetracyanoethylene
Show More Show Less
Hazard Category GHS06: Skull and crossbones
ShelfLife 5 years
EINECSNumber 211-578-0
RTECSNumber KM7300000
Recommended Storage Refrigerator +4°C