accessibility menu, dialog, popup

UserName

CAS RN 66943-05-3

OOOOH2N

CAS RN 66943-05-3

IUPAC Name: 1,4,7,10-tetraoxa-13-azacyclopentadecan-13-ium
Synonyms: 1,4,7,10-tetraoxa-13-azacyclopentadecan-13-ium
Molecular Weight (g/mol): 220.29
Molecular Formula: C10H22NO4
InChi Key: BJUOQSZSDIHZNP-UHFFFAOYSA-O
SMILES: C1COCCOCCOCCOCC[NH2+]1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)