missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-15-crown 5-Ether 98.0+%, TCI America™

Catalog No. A13235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13235G 5 g
A13231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13235G Supplier TCI America Supplier No. A13235G

Chemical Identifiers

CAS 66943-05-3
Molecular Formula C10H21NO4
Molecular Weight (g/mol) 219.281
MDL Number MFCD00075465
InChI Key BJUOQSZSDIHZNP-UHFFFAOYSA-N
Synonym aza-15-crown-5, 1-aza-15-crown 5-ether, 1-aza-15-crown-5, 1-aza-15-crown, 1-aza-4,7,10,13-tetraoxacyclopentadecane
PubChem CID 544820
IUPAC Name 1,4,7,10-tetraoxa-13-azacyclopentadecane
SMILES C1COCCOCCOCCOCCN1

Specifications

Melting Point 46°C
Color White
Quantity 5 g
Formula Weight 219.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aza-15-crown 5-Ether
TSCA No
Recommended Storage Refrigerator